2-Chloro-4,6-dimethylpyrimidine 97%

Warning

AC44255-0250 AC44255-0050
103513-620EA 292 USD
103513-620 103513-618
2-Chloro-4,6-dimethylpyrimidine 97%
2-Chloro-4,6-dimethylpyrimidine
Formula: C₆H₇ClN₂
Boiling Pt: 210…212 °C (760 mmHg)
Melting Pt: 36…38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023199
CAS Number: 4472-44-0

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 34 to 39 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR