2-Chloro-3-methylbenzotrifluoride 95%

Warning

AC45129-0050 AC45129-0010
103464-452EA 144 USD
103464-452 103464-454
2-Chloro-3-methylbenzotrifluoride 95%
2-Chloro-3-methylbenzotrifluoride
Formula: C₈H₆ClF₃
Boiling Pt: 82…84 °C (40mBar)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD11226661
CAS Number: 74483-48-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR