2-Bromo-5-thiazolecarboxylic acid 97%

Warning

Synonyms: 2-Bromothiazole-5-carboxylic acid

AC42976-0050 AC42976-0010
103464-428EA 262 USD
103464-428 103464-426
2-Bromo-5-thiazolecarboxylic acid 97%
2-Bromo-5-thiazolecarboxylic acid
Formula: C₄H₂BrNO₂S
MW: 208.04 g/mol
Melting Pt: 174.5…175.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04115730
CAS Number: 54045-76-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR