2-Bromo-5-formylthiazole 97%

Warning

Synonyms: 2-Bromothiazole-5-carboxaldehyde

AC43101-0050 AC43101-0010
103524-380EA 152 USD
103524-380 103524-382
2-Bromo-5-formylthiazole 97%
2-Bromo-5-formylthiazole
Formula: C₄H₂BrNOS
MW: 192.03 g/mol
Melting Pt: 112…113.5 °C
Storage Temperature: Refrigerator
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 114 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR