2-Bromomalondialdehyde 97+%

Danger

Synonyms: Bromomalonaldehyde2-Bromomalonaldehyde

AC39790-0010 AC39790-0050 AC39790-0250
103464-390EA 35.3 USD
103464-390 103464-394 103464-392
2-Bromomalondialdehyde 97+%
2-Bromomalondialdehyde
Formula: C₃H₃BrO₂
MW: 150.96 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00459999
CAS Number: 2065-75-0

Order Now

Specification Test Results

Appearance White to yellow to brown Powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR