2-Bromo-5-methoxybenzoic acid 97%

Warning

AC39893-0050
103464-338EA 53.2 USD
103464-338
2-Bromo-5-methoxybenzoic acid 97%
2-Bromo-5-methoxybenzoic acid
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00020214
CAS Number: 22921-68-2

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 156 to 160 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR