2-Bromo-5-hydroxypyrazine 97%

Warning

Synonyms: 5-Bromopyrazin-2-ol

AC43589-0010 AC43589-0050
103464-332EA 473 USD
103464-332 103464-330
2-Bromo-5-hydroxypyrazine 97%
2-Bromo-5-hydroxypyrazine
Formula: C₄H₃BrN₂O
MW: 174.9800 g/mol
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD06245330
CAS Number: 374063-92-0

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥98.5 %
Specific optical rotation +67° to +69° (20 °C, 589 nm) (c=2.3, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR