2-Bromo-1-indanol 99%

Synonyms: 2-Bromo-1-hydroxyindane

AC10705-2500 AC10705-0100 AC10705-0500
103464-284EA 31.7 USD
103464-284 103464-288 103464-286
2-Bromo-1-indanol 99%
2-Bromo-1-indanol
Formula: C₉H₉BrO
MW: 213.07 g/mol
Melting Pt: 130…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00003798
CAS Number: 5400-80-6
EINECS: 226-442-6

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder or needles
Infrared spectrum Conforms
Melting point 128 to 132 °C
GC ≥98.5 %
Pore volume 0.70 to 0.85 ml/g
Carbon (C) ≥20.7 %
Volatile matter ≤4 % (110 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR