2-Bromo-4-thiazolecarboxylic acid 98%

Warning

Synonyms: 2-Bromo-1,3-thiazole-4-carboxylic acid2-Bromothiazole-4-carboxylic acid

AC45460-0010 AC45460-0050
103464-272EA 452 USD
103464-272 103464-270
2-Bromo-4-thiazolecarboxylic acid 98%
2-Bromo-4-thiazolecarboxylic acid
Formula: C₄H₂BrNO₂S
MW: 208.04 g/mol
Melting Pt: 204…205.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04115729
CAS Number: 5198-88-9

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR