2-Aminomethyl-1H-imidazole dihydrochloride 98%

Warning

Synonyms: (1H-Imidazol-2-yl)methanamine dihydrochloride

AC45459-0010
103517-542EA 81.85 USD
103517-542
2-Aminomethyl-1H-imidazole dihydrochloride 98%
2-Aminomethyl-1H-imidazole dihydrochloride
Formula: C₄H₉Cl₂N₃
MW: 170.04 g/mol
Melting Pt: 250…251 °C
Storage Temperature: Ambient
MDL Number: MFCD06738779
CAS Number: 22600-77-7

Order Now

Specification Test Results

Appearance White to light yellow to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 %
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR