Cysteamine 95%

Danger

Synonyms: 2-Aminoethanethiol

AC43471-0250 AC43471-0050
103524-436EA 81.7 USD
103524-436 103524-438
Cysteamine 95%
Cysteamine
Formula: C₂H₇NS
MW: 77.15 g/mol
Melting Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008196
CAS Number: 60-23-1
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 94 to 99 °C
Titration Iodimetric ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR