2-Aminoacetophenone hydrochloride 96%
Warning
Synonyms: Phenacylamine hydrochloride
AC10310-0010 AC10310-0050 AC10310-0250
103508-484EA 43.8 USD
103508-484 103508-486 103508-488
2-Aminoacetophenone hydrochloride 96%
2-Aminoacetophenone hydrochloride
Order Now
Specification Test Results
Appearance White to yellow to beige to light brown Powder or crystals
Infrared spectrum Conforms
Titration Argentometric 95.0 to 105.0 %
HPLC ≥98.5 %