2-Amino-5-phenylpyrazine 98%
|
Warning
|
AC45250-0050
AC45250-0010
103517-324EA
379.2
USD
103517-324
103517-322
2-Amino-5-phenylpyrazine 98%
2-Amino-5-phenylpyrazine
Specification Test Results
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 143 to 149 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...