2-Amino-4-phenylphenol 98%

Warning

Synonyms: 3-Amino-4-hydroxybiphenyl3-Amino-[1,1'-biphenyl]-4-ol

AC43993-0010
103516-050EA 44.8 USD
103516-050
2-Amino-4-phenylphenol 98%
2-Amino-4-phenylphenol
Formula: C₁₂H₁₁NO
MW: 185.2200 g/mol
Melting Pt: 208.5…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00059187
CAS Number: 1134-36-7

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 206 to 210 °C (Dec)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR