2-Amino-4'-methoxyacetophenone hydrochloride 97%

Warning

AC42571-0050 AC42571-0010
103514-950EA 43.9 USD
103514-950 103514-952
2-Amino-4'-methoxyacetophenone hydrochloride 97%
2-Amino-4'-methoxyacetophenone hydrochloride
Formula: C₉H₁₂ClNO₂
Melting Pt: 190…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00193078
CAS Number: 3883-94-1

Order Now

Specification Test Results

Appearance White to yellow to tan Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR