2-Amino-4-(4-chlorophenyl)thiazole 98%
|
Warning
|
Synonyms:
4-(4-Chlorophenyl)thiazol-2-amine
AC42668-0010
103512-626EA
28.3
USD
103512-626
2-Amino-4-(4-chlorophenyl)thiazole 98%
2-Amino-4-(4-chlorophenyl)thiazole
Formula:
C₉H₇ClN₂S MW: 210.69 g/mol Melting Pt: 166.5…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047058 CAS Number: 2103-99-3 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 167 to 171 °C |
HPLC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Particle size | ≥45 % (> 75 µm) |
Particle size | ≥70 % (> 32 µm) |
Residue after ignition | ≤0.05 % |
pH | 5 to 7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...