3-Nitroanthranilic acid 97%

Warning

Synonyms: 2-Amino-3-nitrobenzoic acid

AC44318-0100 AC44318-0010
103516-284EA 24.9 USD
103516-284 103516-286
3-Nitroanthranilic acid 97%
3-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00024261
CAS Number: 606-18-8

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 204 to 210 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR