2-Amino-3-chloro-5-(trifluoromethyl)pyridine 97%

Warning

AC43627-0250
103515-784EA 46 USD
103515-784
2-Amino-3-chloro-5-(trifluoromethyl)pyridine 97%
2-Amino-3-chloro-5-(trifluoromethyl)pyridine
Formula: C₆H₄ClF₃N₂
MW: 196.5600 g/mol
Melting Pt: 95…97.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00042154
CAS Number: 79456-26-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR