2-Acetamido-5-bromopyridine 98%

Warning

Synonyms: N-(5-Bromopyridin-2-yl)acetamide

AC44242-0010 AC44242-0050
103513-558EA 24.2 USD
103513-558 103513-560
2-Acetamido-5-bromopyridine 98%
2-Acetamido-5-bromopyridine
Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00468968
CAS Number: 7169-97-3

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 175 to 179 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR