2-(2-(2-Chloroethoxy)ethoxy)ethanol 95%

Warning

AC17227-0500 AC17227-0100
103509-112EA 114 USD
103509-112 103509-110
2-(2-(2-Chloroethoxy)ethoxy)ethanol 95%
2-(2-(2-Chloroethoxy)ethoxy)ethanol
Formula: C₆H₁₃ClO₃
MW: 168.62 g/mol
Boiling Pt: 256…258 °C (760 mmHg)
Density: 1.163 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002874
CAS Number: 5197-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR