2,6-Dichlorophenolindophenol sodium salt hydrate 90+%

Synonyms: Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide hydrateDPIP2,6-Dichlorophenolindophenol sodium salt xhydrateDCIPSodium 2,6-dichloroindophenolate hydrate2,6-Dichloroindophenol, sodium salt hydrate2,6-Dichloroindophenol sodium hydrate2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt hydrateTillmans' reagent hydrate

AC45070-0050 AC45070-0250 AC45070-1000
103466-900EA 527 USD
103466-900 103466-898 103466-896
2,6-Dichlorophenolindophenol sodium salt hydrate 90+%
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula: C₁₂H₆Cl₂NNaO₂·xH₂O
MW: 290.08 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150014
CAS Number: 1266615-56-8
EINECS: 210-640-4

Order Now

Specification Test Results

Appearance Dark green to dark blue-purple Powder
Infrared spectrum Conforms
Loss on drying ≤14 % (120 °C)
Titration with HClO4 ≥90.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR