2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine 97%

Warning

AC45229-0010 AC45229-0050
103517-304EA 653 USD
103517-304 103518-364
2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine 97%
2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine
Formula: C₆HCl₂F₄N
Boiling Pt: 90 °C (55mbar)
Storage Temperature: Ambient
MDL Number: MFCD11100554
CAS Number: 1099598-11-4

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR