2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 95%

Warning

Synonyms: 2,6-Dichloro-7-deazapurine

AC45401-2500 AC45401-0010
103518-386EA 622.6 USD
103518-386 103518-388
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 95%
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₆H₃Cl₂N₃
MW: 188.01 g/mol
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD08059278
CAS Number: 90213-66-4

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR