2,4-Dichloro-6-methylpyridine 97%

Warning

AC45377-0050 AC45377-0010
103517-286EA 124.8 USD
103517-286 103517-288
2,4-Dichloro-6-methylpyridine 97%
2,4-Dichloro-6-methylpyridine
Formula: C₆H₅Cl₂N
MW: 162.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669834
CAS Number: 42779-56-6

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR