2,4-Dichloro-5-nitropyrimidine 97%

Warning

AC43529-0010 AC43529-0250 AC43529-0050
103513-188EA 93.6 USD
103513-188 103515-684 103513-186
2,4-Dichloro-5-nitropyrimidine 97%
2,4-Dichloro-5-nitropyrimidine
Formula: C₄HCl₂N₃O₂
MW: 193.98 g/mol
Boiling Pt: 135 °C (17Torr)
Melting Pt: 29…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00127867
CAS Number: 49845-33-2

Order Now

Specification Test Results

Appearance Clear yellow Liquid or low melting solid
Infrared spectrum Conforms
Melting point 28 to 32 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR