2,1,3-Benzothiadiazole 98%

Warning

AC10566-0050 AC10566-0250
103508-420EA 38.3 USD
103508-420 103508-422
2,1,3-Benzothiadiazole 98%
2,1,3-Benzothiadiazole
Formula: C₆H₄N₂S
Boiling Pt: 206 °C (760 mmHg)
Melting Pt: 42…44 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00005809
CAS Number: 273-13-2

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR