2-Pyrrolidino-5-pyrimidineboronic acid pinacol ester 95%

Warning

Synonyms: 2-(Pyrrolidin-1-yl)pyrimidine-5-boronic acid pinacol ester2-(Pyrrolidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

AC45435-2500 AC45435-0010
103517-478EA 168.7 USD
103517-478 103517-476
2-Pyrrolidino-5-pyrimidineboronic acid pinacol ester 95%
2-Pyrrolidino-5-pyrimidineboronic acid pinacol ester
Formula: C₁₄H₂₂BN₃O₂
MW: 275.1544 g/mol
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD07781249
CAS Number: 1015242-07-5

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR