2-(3-Bromophenyl)pyridine 97%

Warning

AC45756-0050 AC45756-0010
103518-480EA 100.6 USD
103518-480 103518-482
2-(3-Bromophenyl)pyridine 97%
2-(3-Bromophenyl)pyridine
Formula: C₁₁H₈BrN
MW: 234.0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749846
CAS Number: 4373-60-8

Order Now

Specification Test Results

Appearance Yellow to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR