2-(2-Bromoethyl)-1,3-dioxolane 96%

Warning

AC21354-0250 AC21354-0100 AC21354-0500
103526-722EA 159 USD
103526-722 103526-720 103526-718
2-(2-Bromoethyl)-1,3-dioxolane 96%
2-(2-Bromoethyl)-1,3-dioxolane
Formula: C₅H₉BrO₂
Boiling Pt: 68…70 °C (8 mmHg)
Density: 1.54 g/cm³
Flash Pt: 65 °C
Storage Temperature: Freezer
MDL Number: MFCD00003216
CAS Number: 18742-02-4
EINECS: 242-551-1
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4830 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR