1-Octanol, anhydrous 99%, AcroSeal™
|
Warning
|
Synonyms:
n-Octanol
AC43458-0010
AC43458-1000
103515-566EA
38.5
USD
103515-566
103515-564
1-Octanol, anhydrous 99%, AcroSeal™
1-Octanol
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol Boiling Pt: 195 °C (760 mmHg) Melting Pt: –16 °C Density: 0.824 g/cm³ Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 Merck Index: 12,06849 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥98.5 % |
Residue after evaporation | ≤0.004 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...