1-Octanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography

Warning

Synonyms: Sodium 1-octansulfonateSodium octane-1-sulphonate

AC39669-0250 AC39669-1000
103517-930EA 837 USD
103517-930 103517-928
1-Octanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://us.vwr.com/stibo/web/std.lang.all/09/50/2300950.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 % (150 °C)
UV (0.4 M in water)
UV at 210 nm A: ≤0.1
UV at 220 nm A: ≤0.06
UV at 230 nm A: ≤0.04
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.4 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR