1-Naphthyl phosphate disodium salt monohydrate 98%

Warning

Synonyms: 1-Naphthyl phosphate, disodium salt hydrate

AC39826-0010 AC39826-0050 AC39826-0250
103518-812EA 68.5 USD
103518-812 103518-814 103518-816
1-Naphthyl phosphate disodium salt monohydrate 98%
1-Naphthyl phosphate disodium salt monohydrate
Formula: C₁₀H₇Na₂O₄P·1H₂O
MW: 268.12 (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00150614
CAS Number: 207569-06-0

Order Now

Specification Test Results

Appearance White to pale brown Powder
Infrared spectrum Conforms
Water ≤15.0 %
HPLC 97.5 %
Titration with HClO4 97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR