1-Methyl-1H-indole-5-carboxylic acid 95%

Warning

Synonyms: 1-Methyl-5-indolecarboxylic Acid

AC44864-0050 AC44864-0010
103516-848EA 527.1 USD
103516-848 103516-846
1-Methyl-1H-indole-5-carboxylic acid 95%
1-Methyl-1H-indole-5-carboxylic acid
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 221…223 °C
Storage Temperature: Ambient
MDL Number: MFCD03839859
CAS Number: 186129-25-9

Order Now

Specification Test Results

Appearance White to yellow to pink Solid
Infrared spectrum Conforms
Melting point 219 to 225 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR