1-Methyl-1H-indazole-4-boronic acid pinacol ester 97%

Warning

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

AC44513-0050
103513-806EA 819.7 USD
103513-806
1-Methyl-1H-indazole-4-boronic acid pinacol ester 97%
1-Methyl-1H-indazole-4-boronic acid pinacol ester
Formula: C₁₄H₁₉BN₂O₂
MW: 258.1239 g/mol
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD10700158
CAS Number: 885698-94-2

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 82 to 87 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR