1-Methyl-1H-indazole-4-boronic acid 95%

Warning

Synonyms: (1-Methyl-1H-indazol-4-yl)boronic acid

AC45434-0010
103517-472EA 265.1 USD
103517-472
1-Methyl-1H-indazole-4-boronic acid 95%
1-Methyl-1H-indazole-4-boronic acid
Formula: C₈H₉BN₂O₂
MW: 175.9803 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870045
CAS Number: 1001907-60-3

Order Now

Specification Test Results

Appearance White to off-white Powder or solid
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR