1H-Indazole-5-boronic acid 97%

Warning

AC43035-0010
103515-264EA 218.7 USD
103515-264
1H-Indazole-5-boronic acid 97%
1H-Indazole-5-boronic acid
Formula: C₇H₇BN₂O₂
MW: 161.9537 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739151
CAS Number: 338454-14-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤5 ppm
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤5 ppm
Chromium (Cr) ≤0.5 ppm
Iron (Fe) ≤0.5 ppm
Lithium (Li) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤0.5 ppm
Potassium (K) ≤5 ppm
Sodium (Na) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR