1H-Indazole-4-carboxylic acid 97%

Warning

AC43009-0050
103515-144EA 436.8 USD
103515-144
1H-Indazole-4-carboxylic acid 97%
1H-Indazole-4-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 209.5…211 °C
Storage Temperature: Ambient
MDL Number: MFCD03840644
CAS Number: 677306-38-6

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 207 to 212 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR