1-Hexanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography

Warning

Synonyms: Sodium 1-hexanesulfonateSodium hexanesulfonateSodium hexane-1-sulfonateSodium hexanesulphonateSodium 1-hexanesulphonateSodium hexane-1-sulphonate

AC39667-0250 AC39667-1000
103517-922EA 889 USD
103517-922 103517-920
1-Hexanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
1-Hexanesulphonic acid sodium salt
Formula: C₆H₁₃O₃SNa
MW: 188.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://us.vwr.com/stibo/web/std.lang.all/07/94/2300794.jpg

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Loss on drying ≤2 % (150 °C)
UV (0.5 M in water)
UV at 210 nm A: ≤0.1
UV at 220 nm A: ≤0.06
UV at 230 nm A: ≤0.04
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR