1-Bromo-2-(methoxymethyl)benzene 98%

Warning

Synonyms: 1-Bromobenzyl methyl ether

AC39340-0010 AC39340-0050 AC39340-0250
103514-366EA 37.3 USD
103514-366 103514-368 103514-370
1-Bromo-2-(methoxymethyl)benzene 98%
1-Bromo-2-(methoxymethyl)benzene
Formula: C₈H₉BrO
MW: 201.0605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03790887
CAS Number: 52711-30-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5470 to 1.5500 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR