5-Bromo-1,2,3-trimethoxybenzene 97%

Warning

AC45813-0250 AC45813-0050 AC45813-1000
103518-528EA 37.3 USD
103518-528 103518-530 103518-532
5-Bromo-1,2,3-trimethoxybenzene 97%
5-Bromo-1,2,3-trimethoxybenzene
Formula: C₉H₁₁BrO₃
MW: 247.09 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00017169
CAS Number: 2675-79-8

Order Now

Specification Test Results

Appearance White to brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 75 to 83 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR