1-Amino-9-fluorenone 97%

Warning

Synonyms: 1-Amino-9H-fluoren-9-one

AC10374-0010
103508-650EA 360.9 USD
103508-650
1-Amino-9-fluorenone 97%
1-Amino-9-fluorenone
Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 120…120.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00001158
CAS Number: 6344-62-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 122 °C
HPLC ≥96.0 %
GC ≥98.5 %
Acidity ≤0.1 % (as HBr)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR