1-Allylpyrazole 97%

Warning

AC45652-0050 AC45652-0010
103517-502EA 413.9 USD
103517-502 103517-500
1-Allylpyrazole 97%
1-Allylpyrazole
Formula: C₆H₈N₂
Boiling Pt: 157…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00034406
CAS Number: 35852-74-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR