10-Deacetylbaccatin III 99%

Warning

AC29477-0010
103512-130EA 619 USD
103512-130
10-Deacetylbaccatin III 99%
10-Deacetylbaccatin III
Formula: C₂₉H₃₆O₁₀
Melting Pt: 221…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00132913
CAS Number: 32981-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR