1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%

Warning

Synonyms: Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate

AC44501-0050 AC44501-0010
103516-380EA 392 USD
103516-380 103516-378
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₃H₂₁BN₂O₄
MW: 280.1278 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD08706026
CAS Number: 864754-16-5

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Viscous liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR