1,8-Bis(dimethylamino)naphthalene 99%

Danger

Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine

AC16986-0100 AC16986-0500
103526-278EA 134 USD
103526-278 103526-276
1,8-Bis(dimethylamino)naphthalene 99%
1,8-Bis(dimethylamino)naphthalene
Formula: C₁₄H₁₈N₂
MW: 214.31 g/mol
Boiling Pt: 144…145 °C (5hPa)
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003920
CAS Number: 20734-58-1
EINECS: 244-001-6
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR