1,6-Dihydroxynaphthalene 99%

Warning

Synonyms: Naphthalene-1,6-diol

AC26991-1000 AC26991-0250
103511-950EA 132 USD
103511-950 103511-948
1,6-Dihydroxynaphthalene 99%
1,6-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003981
CAS Number: 575-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR