1,4-Dioxane-2,3-diol 98%

Warning

Synonyms: 2,3-Dihydroxy-1,4-dioxane

AC43837-0050
103516-028EA 100 USD
103516-028
1,4-Dioxane-2,3-diol 98%
1,4-Dioxane-2,3-diol
Formula: C₄H₈O₄
MW: 120.10 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00006572
CAS Number: 4845-50-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 39.10 to 41.10 % (C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR