2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde 98%

Warning

Synonyms: 2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde2,3-dihydrobenzo[b][1,4]dioxine-6-carbaldehyde1,4-Benzodioxan-6-carboxaldehyde

AC39821-0010 AC39821-0050
103514-506EA 33.5 USD
103514-506 103514-508
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde 98%
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde
Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 105 °C (20hPa)
Melting Pt: 52…53 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00010092
CAS Number: 29668-44-8

Order Now

Specification Test Results

Appearance Yellow to light beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR