1,3-Dimethyl-1H-pyrazole-5-carboxylic acid 97%

Warning

AC44863-0100 AC44863-0250
103516-844EA 638.5 USD
103516-844 103516-842
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid 97%
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formula: C₆H₈N₂O₂
MW: 140.1400 g/mol
Melting Pt: 209…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00159636
CAS Number: 5744-56-9

Order Now

Specification Test Results

Appearance White to off-white Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR