Bis-trispropane ≥99%

Supplier: ACROS ORGANICS MS

Synonyms: 1,3-Bis[tris(hydroxymethyl)amino]propane2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]1,3-Bis(tris(hydroxymethyl)methylamino)propane

AC20264-0010 AC20264-0250 AC20264-2500 AC20264-1000
103509-660EA 1198.45 USD InStock
103509-660 103509-662 103509-666 103509-664
Bis-trispropane ≥99%
BIS-TRIS propane
Formula: C₁₁H₂₆N₂O₆
MW: 282,34 g/mol
Melting Pt: 164…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00004689
CAS Number: 64431-96-5
EINECS: 264-899-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 161°C to 168°C
Elemental Analysis 46.33 to 47.2 % C
Elemental Analysis 9.82 to 10.02 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR